Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MTF2 Rabbit Polyclonal Antibody
Anti-MTF2 Rabbit Polyclonal Antibody
Catalog # ABCA_AB262914-100U
Supplier:  Abcam
New Product
undefined
Anti-MTF2 Rabbit Polyclonal Antibody
Catalog # ABCA_AB262914-100U
Supplier:  Abcam
Supplier Number:  AB262914-100UL
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    PCL2
  • Antigen Symbol:
    MTF2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Size:
    100 µl
  • Form:
    Liquid
  • Antigen Synonyms:
    Metal-response element-binding transcription factor 2|Metal regulatory transcription factor 2|Metal-response element DNA-binding protein M96|Polycomb-like protein 2|h
  • Molecular Weight:
    67090 Da
  • Storage Temperature:
    Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Recombinant Fragment Protein within Human MTF2 aa 400-550. The exact immunogen used to generate this antibody is proprietary information.
  • Tested Applications:
    ICC/IF
  • Purification:
    Immunogen affinity purification
  • Packaging:
    Vial

Specifications

About this item

Rabbit polyclonal to MTF2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MTF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human