Anti-GRHL2 Mouse Monoclonal Antibody [clone: CL3760]
Catalog # ABCA_AB236758-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:BOM
- Antigen Symbol:GRHL2
- Clonality:Monoclonal
- Clone:CL3760
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Size:100 µl
- Epitope:Binds to an epitope located within the peptide sequence VVKAEDFTPV as determined by overlapping synthetic peptides
- Form:Liquid
- Antigen Synonyms:TFCP2L3|Grainyhead-like protein 2 homolog|Brother of mammalian grainyhead|Transcription factor CP2-like 3
- Molecular Weight:71105 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human GRHL2 aa 100-300. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC, IHC-P
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [CL3760] to GRHL2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GRHL2
Clonality: Monoclonal
Clone: CL3760
Conjugation:
Epitope: Binds to an epitope located within the peptide sequence VVKAEDFTPV as determined by overlapping synthetic peptides
Host: Mouse
Isotype: IgG1
Reactivity: Human