Anti-Dengue Virus NS1 glycoprotein Mouse Monoclonal Antibody [clone: JE1.C10.H2.A9]
Catalog # ABCA_AB253231-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Genome polyprotein
- Antigen Symbol:Dengue Virus NS1 glycoprotein
- Clonality:Monoclonal
- Clone:JE1.C10.H2.A9
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Size:100 µg
- Form:Liquid
- Molecular Weight:379547 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Concentration:1.5 mg/ml
- Shipping Temperature:4 °C
- Purification:Protein G Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [JE1.C10.H2.A9] to Dengue Virus NS1 glycoprotein.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Control
Antigen: IgG
Clonality: Monoclonal
Clone: PIP
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian hamster
Isotype: IgG
Reactivity: