Anti-Oct4 Rabbit Monoclonal Antibody [clone: EPR17929] (PE (Phycoerythrin))
ABCA_AB283731-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:OCT3
- Antigen Symbol:Oct4
- Clonality:Monoclonal
- Clone:EPR17929
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:OTF3|POU5F1|Octamer-binding protein 3|Octamer-binding protein 4|Octamer-binding transcription factor 3|45933OTF-3
- Molecular Weight:38571 Da
- Storage Temperature:Store at 4 °C (stable for up to 12 months). Upon delivery aliquot. Store at 4 °C. Store in the dark.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested Applications:Flow Cyt (Intra)
- Purification:Protein A affinity purification
- Packaging:Vial
PE Rabbit monoclonal [EPR17929] to Oct4.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Oct4
Clonality: Monoclonal
Clone: EPR17929
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human