Anti-LAMP1 Mouse Monoclonal Antibody [clone: H4A3]
Catalog # ABCA_AB25630-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CD107a
- Antigen Symbol:LAMP1
- Clonality:Monoclonal
- Clone:H4A3
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Lysosome-associated membrane glycoprotein 1|LAMP-1|Lysosome-associated membrane protein 1|CD107 antigen-like family member A
- Storage Buffer:Borate buffered saline
- Molecular Weight:44882 Da
- Storage Temperature:Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:0.1 mg/ml
- Shipping Temperature:4 °C
- Tested Applications:Flow Cyt (Intra), ICC/IF, IHC-P
- Purification:Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [H4A3] to LAMP1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: H4A3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human