Anti-Melanoma gp100 Mouse Monoclonal Antibody [clone: HMB45]
Catalog # ABCA_AB787-100UG
Supplier: Abcam
New Product
Specifications
- Size:100 µg
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [HMB45] to Melanoma gp100.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Melanoma gp100
Clonality: Monoclonal
Clone: HMB45
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human