Anti-Adiponectin Rabbit Monoclonal Antibody [clone: EPR21976-244]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR21976-244] to Adiponectin - BSA and Azide free (Detector).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Adiponectin
Clonality: Monoclonal
Clone: EPR21976-244
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Specifications
- Catalog No:
- ABCA_AB245079-100U
- ABCA_AB245079-1MG
- Antigen Symbol:
- Adiponectin
- Adiponectin
- Antigen Name:
- 30 kDa adipocyte complement-related protein
- 30 kDa adipocyte complement-related protein
- Antigen Synonyms:
- ACDC|ADIPO_HUMAN|ADIPQTL1|ADPN|Acrp 30|Adipocyte|Adipocyte C1q and collagen domain containing protein|Adipocyte complement related protein of 30 kDa|Adipocyte complement-related 30 kDa protein|Adiponectin precursor|Adipose most abundant gene transcript 1|Adipose most abundant gene transcript 1 protein|Adipose specific collagen like factor|C1q and collagen domain-containing protein|GBP 28|Gelatin-binding protein|OTTHUMP00000210047|adipocyte-specific secretory protein|C1Q and collagen domain containing|apM-1|gelatin-binding protein 28
- ACDC|ADIPO_HUMAN|ADIPQTL1|ADPN|Acrp 30|Adipocyte|Adipocyte C1q and collagen domain containing protein|Adipocyte complement related protein of 30 kDa|Adipocyte complement-related 30 kDa protein|Adiponectin precursor|Adipose most abundant gene transcript 1|Adipose most abundant gene transcript 1 protein|Adipose specific collagen like factor|C1q and collagen domain-containing protein|GBP 28|Gelatin-binding protein|OTTHUMP00000210047|adipocyte-specific secretory protein|C1Q and collagen domain containing|apM-1|gelatin-binding protein 28
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR21976-244
- EPR21976-244
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- sELISA
- sELISA
Specifications
Frequently Bought Together