Anti-LAT2 Mouse Monoclonal Antibody [clone: UMAB70]
Catalog # ABCA_AB273674-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:SLC7A8
- Antigen Symbol:LAT2
- Clonality:Monoclonal
- Clone:UMAB70
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- ChIP:Yes
- Form:Liquid
- Antigen Synonyms:Large neutral amino acids transporter small subunit 2|L-type amino acid transporter 2|Solute carrier family 7 member 8|hLAT2
- Molecular Weight:58382 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human SLC7A8.
- Tested Applications:ICC/IF, IHC-P, Protein Microarray ChIP
- Purification:Protein A/G Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [UMAB70] to LAT2 - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: LAT2
Clonality: Monoclonal
Clone: UMAB70
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human