Anti-MIP-1 alpha/CCL3+CCL3L1 Rabbit Monoclonal Antibody [clone: EPR22529-19] (PE (Phycoerythrin))
Supplier: Abcam
New Product
Certificates
About this item
PE Rabbit monoclonal [EPR22529-19] to MIP-1 alpha/CCL3 + CCL3L1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MIP-1 alpha/CCL3+CCL3L1
Clonality: Monoclonal
Clone: EPR22529-19
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB317071-100U
- ABCA_AB317071-10UL
- Antigen Symbol:
- MIP-1 alpha/CCL3+CCL3L1
- MIP-1 alpha/CCL3+CCL3L1
- Antigen Name:
- G0S19-1
- G0S19-1
- Antigen Synonyms:
- MIP1A|SCYA3|CCL3|C-C motif chemokine 3|G0/G1 switch regulatory protein 19-1|Macrophage inflammatory protein 1-alpha|PAT 464.1|SIS-beta|Small-inducible cytokine A3|Tonsillar lymphocyte LD78 alpha protein|MIP-1-alpha
- MIP1A|SCYA3|CCL3|C-C motif chemokine 3|G0/G1 switch regulatory protein 19-1|Macrophage inflammatory protein 1-alpha|PAT 464.1|SIS-beta|Small-inducible cytokine A3|Tonsillar lymphocyte LD78 alpha protein|MIP-1-alpha
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- EPR22529-19
- EPR22529-19
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 10085 Da
- 10085 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at 4 °C. Avoid freeze/thaw cycle. Store in the dark.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at 4 °C. Avoid freeze/thaw cycle. Store in the dark.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Flow Cyt (Intra)
- Flow Cyt (Intra)