Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-LXR beta/NER Rabbit Polyclonal Antibody
Catalog # ABCA_AB28479-100UG
Supplier:  Abcam
New Product
undefined
Anti-LXR beta/NER Rabbit Polyclonal Antibody
Catalog # ABCA_AB28479-100UG
Supplier:  Abcam
Supplier Number:  AB28479-100UG
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Lxrb
  • Antigen Symbol:
    LXR beta/NER
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Liquid
  • Antigen Synonyms:
    Nr1h2|Oxysterols receptor LXR-beta|Liver X receptor beta|Nuclear receptor subfamily 1 group H member 2|Orphan nuclear receptor OR-1|Ubiquitously-expressed nuclear receptor|UR
  • Storage Buffer:
    PBS, BSA
  • Storage Temperature:
    Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping Temperature:
    Shipped at +4 °C
  • Immunogen:
    The exact immunogen used to generate this antibody is proprietary information.
  • Tested Applications:
    ICC/IF
  • Purification:
    Immunogen Affinity purification
  • Packaging:
    Vial

Specifications

About this item

Rabbit polyclonal to LXR beta/NER.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LXR beta/NER
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human