Anti-S100A9 Rat Monoclonal Antibody [clone: 2B10]
Catalog # ABCA_AB105472-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CAGB
- Antigen Symbol:S100A9
- Clonality:Monoclonal
- Clone:2B10
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:CFAG|MRP14|Protein S100-A9|Calgranulin-B|Calprotectin L1H subunit|Leukocyte L1 complex heavy chain|Migration inhibitory factor-related protein 14|S100 calcium-binding protein A9|MRP-14|p14
- Storage Buffer:PBS
- Molecular Weight:13242 Da
- Storage Temperature:Upon delivery aliquot and store at −20 or −80 °C. Avoid repeated freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Mouse S100a9
- Tested Applications:ICC/IF, IHC-P
- Purification:Protein G affinity purification
- Packaging:Vial
Specifications
About this item
Rat monoclonal [2B10] to S100A9.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: S100A9
Clonality: Monoclonal
Clone: 2B10
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse;Human