Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-Histone H2B (formyl K116) Rabbit Monoclonal Antibody [clone: EPR17699]
 :  Abcam
New Product
 
Anti-Histone H2B (formyl K116) Rabbit Monoclonal Antibody [clone: EPR17699]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCA_AB177786-1ML
  • ABCA_AB177786-100U
  • Antigen Symbol:
  • Histone H2B
  • Histone H2B
  • Antigen Name:
  • H2BFQ
  • H2BFQ
  • Antigen Synonyms:
  • HIST2H2BE|H2BC21|Histone H2B type 2-E|H2B-clustered histone 21|Histone H2B-GL105|Histone H2B.q|H2B/q|H2BK116formyl
  • HIST2H2BE|H2BC21|Histone H2B type 2-E|H2B-clustered histone 21|Histone H2B-GL105|Histone H2B.q|H2B/q|H2BK116formyl
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR17699
  • EPR17699
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Modification:
  • formyl K116
  • formyl K116
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 13920 Da
  • 13920 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • Tested Applications:
  • ICC/IF, PepArr
  • ICC/IF, PepArr