Anti-PGP9.5 Rabbit Monoclonal Antibody [clone: EPR4118] (Alexa Fluor® 594)
Supplier: Abcam
New Product
Certificates
About this item
Alexa Fluor® 594 Rabbit monoclonal [EPR4118] to PGP9.5 - Neuronal Marker.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PGP9.5
Clonality: Monoclonal
Clone: EPR4118
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Specifications
- Catalog No:
- ABCA_AB302664-10UL
- ABCA_AB302664-100U
- Antigen Symbol:
- PGP9.5
- PGP9.5
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase isozyme L1
- Ubiquitin carboxyl-terminal hydrolase isozyme L1
- Antigen Synonyms:
- UCH-L1|Neuron cytoplasmic protein 9.5|PGP 9.5|Ubiquitin thioesterase L1|UCHL1
- UCH-L1|Neuron cytoplasmic protein 9.5|PGP 9.5|Ubiquitin thioesterase L1|UCHL1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 594
- Alexa Fluor® 594
- Clone:
- EPR4118
- EPR4118
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 24824 Da
- 24824 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle. Store in the dark.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle. Store in the dark.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- ICC/IF
- ICC/IF
Specifications
Frequently Bought Together