Anti-SLC27A5/BAL Mouse Monoclonal Antibody [clone: CL0213]
Catalog # ABCA_AB154140-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:ACSB
- Antigen Symbol:SLC27A5/BAL
- Clonality:Monoclonal
- Clone:CL0213
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:ACSVL6|FACVL3|FATP5|SLC27A5|Long-chain fatty acid transport protein 5|FATP-5|Fatty acid transport protein 5|Bile acid-CoA ligase|Bile acyl-CoA synthetase|Cholate--CoA ligase|Long-chain-fatty-acid--CoA ligase|Solute carrier family 27 member 5|Very long-chain acyl-CoA synthetase homolog 2|Very long-chain acyl-CoA synthetase-related protein|BA-CoA ligase|BAL|BACS|VLCS-H2|VLCSH2|VLACS-related|VLACSR
- Molecular Weight:75385 Da
- Storage Temperature:Upon delivery aliquot and store at −20 °C. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human SLC27A5 aa 200-350. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [CL0213] to SLC27A5/BAL.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SLC27A5/BAL
Clonality: Monoclonal
Clone: CL0213
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human