Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: VIPL2] (FITC (Fluorescein Isothiocyanate))
Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: VIPL2] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCA_AB269291-100T
Supplier:  Abcam
New Product
Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: VIPL2] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCA_AB269291-100T
Supplier:  Abcam
Supplier Number:  AB269291-100TEST
New Product

About this item

FITC Mouse monoclonal [VIPL2] to Integrin beta 3.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: VIPL2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human