Anti-Chk2 (phospho T68) Rabbit Monoclonal Antibody [clone: Chk2T68-D12]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [Chk2T68-D12] to Chk2 (phospho T68).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Chk2
Clonality: Monoclonal
Clone: Chk2T68-D12
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- ABCA_AB278548-20UL
- ABCA_AB278548-100U
- Antigen Symbol:
- Chk2
- Chk2
- Antigen Name:
- CDS1
- CDS1
- Antigen Synonyms:
- RAD53|CHEK2|Serine/threonine-protein kinase Chk2|CHK2 checkpoint homolog|Cds1 homolog|Checkpoint kinase 2|HuhCds1
- RAD53|CHEK2|Serine/threonine-protein kinase Chk2|CHK2 checkpoint homolog|Cds1 homolog|Checkpoint kinase 2|HuhCds1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- Chk2T68-D12
- Chk2T68-D12
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Modification:
- phospho T68
- phospho T68
- Immunogen:
- Synthetic Peptide within Human CHEK2 phospho T68. The exact immunogen used to generate this antibody is proprietary information.
- Synthetic Peptide within Human CHEK2 phospho T68. The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 60915 Da
- 60915 Da
- Purification:
- Protein A/G affinity purification
- Protein A/G affinity purification
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C