Anti-RPL26+RPL26L1 Rabbit Polyclonal Antibody
ABCA_AB59567-50UL
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:Large ribosomal subunit protein uL24
- Antigen Symbol:RPL26+RPL26L1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:60S ribosomal protein L26|RPL26
- Storage Buffer:PBS
- Molecular Weight:17258 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human RPL26 aa 1-50 conjugated to Keyhole Limpet Haemocyanin. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to RPL26 + RPL26L1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RPL26+RPL26L1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat