Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Cytochrome P450 17A1/CYP17A1 Rabbit Polyclonal Antibody
Anti-Cytochrome P450 17A1/CYP17A1 Rabbit Polyclonal Antibody
  ABCA_AB231914-100U
 :  Abcam
New Product
Anti-Cytochrome P450 17A1/CYP17A1 Rabbit Polyclonal Antibody
  ABCA_AB231914-100U
 :  Abcam
 :  AB231914-100UG
New Product

 

  • Size:
    100 µg
  • Packaging:
    Vial

 

 

Rabbit polyclonal to Cytochrome P450 17A1/CYP17A1.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Cytochrome P450 17A1/CYP17A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat