Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Melatonin Receptor 1A/MTNR1A Goat Polyclonal Antibody
Anti-Melatonin Receptor 1A/MTNR1A Goat Polyclonal Antibody
Catalog # ABCA_AB87639-200UL
Supplier:  Abcam
New Product
Anti-Melatonin Receptor 1A/MTNR1A Goat Polyclonal Antibody
Catalog # ABCA_AB87639-200UL
Supplier:  Abcam
Supplier Number:  AB87639-200UL
New Product

Specifications

  • Size:
    200 µl
  • Packaging:
    Vial

Specifications

About this item

Goat polyclonal to Melatonin Receptor 1A/MTNR1A.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Melatonin Receptor 1A/MTNR1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat