Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Leukotriene B4 Receptor/BLT Rabbit Monoclonal Antibody [clone: EPR7113]
Anti-Leukotriene B4 Receptor/BLT Rabbit Monoclonal Antibody [clone: EPR7113]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-Leukotriene B4 Receptor/BLT Rabbit Monoclonal Antibody [clone: EPR7113]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB131041-1ML
  • ABCA_AB131041-100U
  • Antigen Symbol:
  • Leukotriene B4 Receptor/BLT
  • Leukotriene B4 Receptor/BLT
  • Antigen Name:
  • BLT
  • BLT
  • Antigen Synonyms:
  • BLT1|BLTR|CMKRL1|GPR16|P2RY7|LTB4R|Leukotriene B4 receptor 1|LTB4-R 1|LTB4-R1|Chemoattractant receptor-like 1|G-protein coupled receptor 16|P2Y purinoceptor 7|P2Y7
  • BLT1|BLTR|CMKRL1|GPR16|P2RY7|LTB4R|Leukotriene B4 receptor 1|LTB4-R 1|LTB4-R1|Chemoattractant receptor-like 1|G-protein coupled receptor 16|P2Y purinoceptor 7|P2Y7
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR7113
  • EPR7113
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 37557 Da
  • 37557 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at −20 °C. Stable for 12 months at −20 °C.
  • Store at −20 °C. Stable for 12 months at −20 °C.
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • Tested Applications:
  • IHC-P
  • IHC-P

Specifications