Anti-Smad1+Smad2+Smad3+SMAD5 (phospho S423 + S425) Rabbit Monoclonal Antibody [clone: EP823Y]
: Abcam
New Product
Rabbit monoclonal [EP823Y] to Smad1 (phospho S463 + S465) + Smad2 (phospho S465 + S467) + Smad3 (phospho S423 + S425) + SMAD5 (phospho S463 + S465).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Smad1+Smad2+Smad3+SMAD5
Clonality: Monoclonal
Clone: EP823Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
- Catalog No:
- ABCA_AB52903-1ML
- ABCA_AB52903-20UL
- ABCA_AB52903-100UL
- Antigen Symbol:
- Smad1+Smad2+Smad3+SMAD5
- Smad1+Smad2+Smad3+SMAD5
- Smad1+Smad2+Smad3+SMAD5
- Antigen Name:
- MADH3
- MADH3
- MADH3
- Antigen Synonyms:
- SMAD3|Mothers against decapentaplegic homolog 3|MAD homolog 3|Mad3|Mothers against DPP homolog 3|hMAD-3|JV15-2|SMAD family member 3|SMAD 3|hSMAD3
- SMAD3|Mothers against decapentaplegic homolog 3|MAD homolog 3|Mad3|Mothers against DPP homolog 3|hMAD-3|JV15-2|SMAD family member 3|SMAD 3|hSMAD3
- SMAD3|Mothers against decapentaplegic homolog 3|MAD homolog 3|Mad3|Mothers against DPP homolog 3|hMAD-3|JV15-2|SMAD family member 3|SMAD 3|hSMAD3
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- EP823Y
- EP823Y
- EP823Y
- Reactivity:
- Mouse, Human
- Mouse, Human
- Mouse, Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Modification:
- phospho S423 + S425
- phospho S423 + S425
- phospho S423 + S425
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 48081 Da
- 48081 Da
- 48081 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Storage Buffer:
- BSA, glycerol, sodium chloride, tris glycine
- BSA, glycerol, sodium chloride, tris glycine
- BSA, glycerol, sodium chloride, tris glycine
- Storage Temperature:
- Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- 4 °C
- 4 °C
- 4 °C
- Tested Applications:
- ChIC/CUT&RUN-seq, Dot, ICC/IF, IHC-P
- ChIC/CUT&RUN-seq, Dot, ICC/IF, IHC-P
- ChIC/CUT&RUN-seq, Dot, ICC/IF, IHC-P