Anti-CD4 Rabbit Recombinant Multiclonal Antibody [clone: RM1136]
: Abcam
New Product
Rabbit recombinant multiclonal [RM1136] to CD4.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD4
Clonality: Recombinant Multiclonal
Clone: RM1136
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- ABCA_AB316866-1ML
- ABCA_AB316866-100U
- ABCA_AB316866-20UL
- Antigen Symbol:
- CD4
- CD4
- CD4
- Antigen Name:
- T-cell surface glycoprotein CD4
- T-cell surface glycoprotein CD4
- T-cell surface glycoprotein CD4
- Antigen Synonyms:
- T-cell surface antigen T4/Leu-3
- T-cell surface antigen T4/Leu-3
- T-cell surface antigen T4/Leu-3
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Recombinant Multiclonal
- Recombinant Multiclonal
- Recombinant Multiclonal
- Clone:
- RM1136
- RM1136
- RM1136
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 51111 Da
- 51111 Da
- 51111 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- 4 °C
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P
- ICC/IF, IHC-P