Anti-FKBP12 Mouse Monoclonal Antibody [clone: 1E5-A12]
ABCA_AB58072-100UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:FKBP1
- Antigen Symbol:FKBP12
- Clonality:Monoclonal
- Clone:1E5-A12
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:FKBP1A|Peptidyl-prolyl cis-trans isomerase FKBP1A|PPIase FKBP1A|12 kDa FK506-binding protein|Calstabin-1|FK506-binding protein 1A|Immunophilin FKBP12|Rotamase|12 kDa FKBP|FKBP-12|FKBP-1A
- Storage Buffer:PBS
- Molecular Weight:11951 Da
- Storage Temperature:Upon delivery aliquot and store at −20 or −80 °C. Avoid repeated freeze/thaw cycles.
- Concentration:0.36 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human FKBP1A
- Purification:Tissue culture supernatant
- Packaging:Vial
Mouse monoclonal [1E5-A12] to FKBP12.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: FKBP12
Clonality: Monoclonal
Clone: 1E5-A12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human