Anti-CD8 alpha Rat Monoclonal Antibody [clone: 53-6.7] (Biotin)
ABCA_AB25176-100UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:CD8a
- Antigen Symbol:CD8 alpha
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:Biotin
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Lyt-2|Lyt2|T-cell surface glycoprotein CD8 alpha chain|T-cell surface glycoprotein Lyt-2
- Storage Buffer:PBS
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Store at 4 °C. Avoid freeze/thaw cycle. Do not freeze.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Tissue preparation containing Cd8a protein. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Affinity purification
- Packaging:Vial
Biotin Rat monoclonal [53-6.7] to CD8 alpha.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD8 alpha
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse