Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-IRF8 Rabbit Monoclonal Antibody [clone: EPR26381-181]
 :  Abcam
New Product
 
undefined
Anti-IRF8 Rabbit Monoclonal Antibody [clone: EPR26381-181]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCA_AB307293-1MG
  • ABCA_AB307293-100U
  • Antigen Symbol:
  • IRF8
  • IRF8
  • Antigen Name:
  • Icsbp
  • Icsbp
  • Antigen Synonyms:
  • Icsbp1|Interferon regulatory factor 8|IRF-8|Interferon consensus sequence-binding protein
  • Icsbp1|Interferon regulatory factor 8|IRF-8|Interferon consensus sequence-binding protein
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR26381-181
  • EPR26381-181
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 48237 Da
  • 48237 Da
  • Purification:
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Storage Temperature:
  • Store at 4 °C.
  • Store at 4 °C.
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • Tested Applications:
  • Flow Cyt (Intra), ICC/IF, IHC-P
  • Flow Cyt (Intra), ICC/IF, IHC-P

 

Frequently Bought Together