Anti-PBRM1/BAF180 Rabbit Monoclonal Antibody [clone: BL-39-2C3]
Catalog # ABCA_AB243876-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:BAF180
- Antigen Symbol:PBRM1/BAF180
- Clonality:Monoclonal
- Clone:BL-39-2C3
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Format:Whole Molecule
- Form:Liquid
- Antigen Synonyms:PB1|PBRM1|Protein polybromo-1|hPB1|BRG1-associated factor 180|Polybromo-1D
- Molecular Weight:192948 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Recombinant Fragment Protein within Human PBRM1 aa 1 - 300. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC, IHC-P
- Packaging:Vial
Specifications
About this item
Rabbit monoclonal [BL-39-2C3] to PBRM1/BAF180.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PBRM1/BAF180
Clonality: Monoclonal
Clone: BL-39-2C3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse