Anti-TPOR/MPL Rabbit Monoclonal Antibody [clone: S03-1C9]
Catalog # ABCA_AB317404-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CD110
- Antigen Symbol:TPOR/MPL
- Clonality:Monoclonal
- Clone:S03-1C9
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:TPOR|MPL|Thrombopoietin receptor|TPO-R|Myeloproliferative leukemia protein|Proto-oncogene c-Mpl
- Molecular Weight:71245 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Concentration:0.3 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human MPL.
- Purification:Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit monoclonal [S03-1C9] to TPOR/MPL.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TPOR/MPL
Clonality: Monoclonal
Clone: S03-1C9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse