Anti-Galectin 1 Rabbit Polyclonal Antibody
ABCA_AB154351-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:Galectin-1
- Antigen Symbol:Galectin 1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Gal-1|14 kDa laminin-binding protein|14 kDa lectin|Beta-galactoside-binding lectin L-14-I|Galaptin|HBL|HPL|Lactose-binding lectin 1|Lectin galactoside-binding soluble 1|Putative MAPK-activating protein PM12|S-Lac lectin 1|HLBP14|LGALS1
- Molecular Weight:14716 Da
- Storage Temperature:Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human LGALS1 aa 1 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to Galectin 1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Galectin 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse