Anti-RPS3 Rabbit Monoclonal Antibody [clone: EPR7808]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR7808] to RPS3 - Ribosome Marker.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RPS3
Clonality: Monoclonal
Clone: EPR7808
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- ABCA_AB128995-100U
- ABCA_AB128995-1ML
- Antigen Symbol:
- RPS3
- RPS3
- Antigen Name:
- OK/SW-cl.26
- OK/SW-cl.26
- Antigen Synonyms:
- Small ribosomal subunit protein uS3|40S ribosomal protein S3
- Small ribosomal subunit protein uS3|40S ribosomal protein S3
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR7808
- EPR7808
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 26688 Da
- 26688 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at −20 °C. Stable for 12 months at −20 °C.
- Store at −20 °C. Stable for 12 months at −20 °C.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P