Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-PRX Rabbit Monoclonal Antibody [clone: EPR24150-36]
 :  Abcam
New Product
 
undefined
Anti-PRX Rabbit Monoclonal Antibody [clone: EPR24150-36]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCA_AB259953-100U
  • ABCA_AB259953-1ML
  • Antigen Symbol:
  • PRX
  • PRX
  • Antigen Name:
  • KIAA1620
  • KIAA1620
  • Antigen Synonyms:
  • Periaxin
  • Periaxin
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR24150-36
  • EPR24150-36
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 154905 Da
  • 154905 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • Tested Applications:
  • IHC-Fr, IHC-P
  • IHC-Fr, IHC-P