Anti-FAM108C1 Rabbit Polyclonal Antibody
ABCA_AB151040-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:ABHD17C
- Antigen Symbol:FAM108C1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Alpha/beta hydrolase domain-containing protein 17C|Abhydrolase domain-containing protein 17C
- Molecular Weight:35831 Da
- Storage Temperature:Upon delivery aliquot and store at −20 °C. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human ABHD17C aa 100-150. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to FAM108C1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: FAM108C1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human