Anti-CCDC112 Rabbit Polyclonal Antibody
ABCA_AB237742-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:MBC1
- Antigen Symbol:CCDC112
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Coiled-coil domain-containing protein 112|Mutated in bladder cancer protein 1
- Molecular Weight:53565 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human CCDC112 aa 200 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Protein G affinity purification
- Packaging:Vial
Rabbit polyclonal to CCDC112.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CCDC112
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human