Anti-Aquaporin 2 (phospho S256) Rabbit Polyclonal Antibody
Catalog # ABCA_AB111346-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Aquaporin-2
- Antigen Symbol:Aquaporin 2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:AQP-2|ADH water channel|Aquaporin-CD|Collecting duct water channel protein|WCH-CD|Water channel protein for renal collecting duct|AQP-CD|AQP2
- Modification:phospho S256
- Molecular Weight:28837 Da
- Storage Temperature:Store at −20 °C. Stable for 12 months at −20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human Aquaporin-2 phospho S256. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Immunogen affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to Aquaporin 2 (phospho S256).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Aquaporin 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human