Anti-Methionine Aminopeptidase 2/p67 Rabbit Monoclonal Antibody [clone: EPR6886(B)]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR6886(B)] to Methionine Aminopeptidase 2/p67.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Methionine Aminopeptidase 2/p67
Clonality: Monoclonal
Clone: EPR6886(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Specifications
- Catalog No:
- ABCA_AB124953-100U
- ABCA_AB124953-1ML
- Antigen Symbol:
- Methionine Aminopeptidase 2/p67
- Methionine Aminopeptidase 2/p67
- Antigen Name:
- MNPEP
- MNPEP
- Antigen Synonyms:
- P67EIF2|METAP2|Methionine aminopeptidase 2|MAP 2|MetAP 2|Initiation factor 2-associated 67 kDa glycoprotein|Peptidase M|p67
- P67EIF2|METAP2|Methionine aminopeptidase 2|MAP 2|MetAP 2|Initiation factor 2-associated 67 kDa glycoprotein|Peptidase M|p67
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR6886(B)
- EPR6886(B)
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 52892 Da
- 52892 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at −20 °C. Stable for 12 months at −20 °C.
- Store at −20 °C. Stable for 12 months at −20 °C.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Flow Cyt (Intra)
- Flow Cyt (Intra)