Anti-RIAM Rabbit Monoclonal Antibody [clone: EPR2806]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR2806] to RIAM.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RIAM
Clonality: Monoclonal
Clone: EPR2806
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Specifications
- Catalog No:
- ABCA_AB92537-1ML
- ABCA_AB92537-100UL
- Antigen Symbol:
- RIAM
- RIAM
- Antigen Name:
- PREL1
- PREL1
- Antigen Synonyms:
- RARP1|APBB1IP|Amyloid beta A4 precursor protein-binding family B member 1-interacting protein|APBB1-interacting protein 1|Proline-rich EVH1 ligand 1|Proline-rich protein 73|Rap1-GTP-interacting adapter molecule|Retinoic acid-responsive proline-rich protein 1|PREL-1|RARP-1
- RARP1|APBB1IP|Amyloid beta A4 precursor protein-binding family B member 1-interacting protein|APBB1-interacting protein 1|Proline-rich EVH1 ligand 1|Proline-rich protein 73|Rap1-GTP-interacting adapter molecule|Retinoic acid-responsive proline-rich protein 1|PREL-1|RARP-1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR2806
- EPR2806
- Reactivity:
- Mouse, Human
- Mouse, Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 73183 Da
- 73183 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Buffer:
- BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- Storage Temperature:
- Store at −20 °C. Stable for 12 months at −20 °C.
- Store at −20 °C. Stable for 12 months at −20 °C.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P