Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Angiomotin like 1 Rabbit Monoclonal Antibody [clone: EPR11803-97]
Anti-Angiomotin like 1 Rabbit Monoclonal Antibody [clone: EPR11803-97]
Supplier:  Abcam
New Product
Certificates
Anti-Angiomotin like 1 Rabbit Monoclonal Antibody [clone: EPR11803-97]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB171976-1ML
  • ABCA_AB171976-100U
  • Antigen Symbol:
  • Angiomotin like 1
  • Angiomotin like 1
  • Antigen Name:
  • Angiomotin-like protein 1
  • Angiomotin-like protein 1
  • Antigen Synonyms:
  • AMOTL1
  • AMOTL1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR11803-97
  • EPR11803-97
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 106574 Da
  • 106574 Da
  • Purification:
  • Tissue culture supernatant
  • Tissue culture supernatant
  • Storage Temperature:
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • 4 °C
  • 4 °C

Specifications