Anti-CDNF Mouse Monoclonal Antibody [clone: 6G5]
ABCA_AB205551-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:ARMETL1
- Antigen Symbol:CDNF
- Clonality:Monoclonal
- Clone:6G5
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Cerebral dopamine neurotrophic factor|ARMET-like protein 1|Conserved dopamine neurotrophic factor
- Molecular Weight:20964 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human CDNF
- Tested Applications:ICC/IF, IHC-P
- Purification:Protein G affinity purification
- Packaging:Vial
Mouse monoclonal [6G5] to CDNF.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CDNF
Clonality: Monoclonal
Clone: 6G5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human