Anti-Histone H3 (mono methyl K4) Rabbit Monoclonal Antibody [clone: ERP16597]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [ERP16597] to Histone H3 (mono methyl K4) - ChIP Grade.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: ERP16597
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Specifications
- Catalog No:
- ABCA_AB176877-100U
- ABCA_AB176877-1ML
- Antigen Symbol:
- Histone H3
- Histone H3
- Antigen Name:
- H3FA
- H3FA
- Antigen Synonyms:
- HIST1H3A|H3C2|H3FL|HIST1H3B|H3C3|H3FC HIST1H3C|H3C4|H3FB|HIST1H3D|H3C6|H3FD|HIST1H3E|H3C7|H3FI|HIST1H3F|H3C8|H3FH|HIST1H3G|H3C10|H3FK|HIST1H3H|H3C11|H3FF|HIST1H3I|H3C12|H3FJ|HIST1H3J|H3FC|HIST1H3C|H3C1|Histone H3.1|Histone H3/a|Histone H3/b|Histone H3/c|Histone H3/d|Histone H3/f|Histone H3/h|Histone H3/i|Histone H3/j|Histone H3/k|Histone H3/l|H3K4me
- HIST1H3A|H3C2|H3FL|HIST1H3B|H3C3|H3FC HIST1H3C|H3C4|H3FB|HIST1H3D|H3C6|H3FD|HIST1H3E|H3C7|H3FI|HIST1H3F|H3C8|H3FH|HIST1H3G|H3C10|H3FK|HIST1H3H|H3C11|H3FF|HIST1H3I|H3C12|H3FJ|HIST1H3J|H3FC|HIST1H3C|H3C1|Histone H3.1|Histone H3/a|Histone H3/b|Histone H3/c|Histone H3/d|Histone H3/f|Histone H3/h|Histone H3/i|Histone H3/j|Histone H3/k|Histone H3/l|H3K4me
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- ERP16597
- ERP16597
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Modification:
- mono methyl K4
- mono methyl K4
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 15404 Da
- 15404 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- ChIC/CUT&RUN-seq, ChIP-seq, ICC/IF, IHC-P, PepArr
- ChIC/CUT&RUN-seq, ChIP-seq, ICC/IF, IHC-P, PepArr