Anti-Desmoglein 3/PVA Rabbit Monoclonal Antibody [clone: EPR14101]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR14101] to Desmoglein 3/PVA.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Desmoglein 3/PVA
Clonality: Monoclonal
Clone: EPR14101
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB183743-1ML
- ABCA_AB183743-100U
- Antigen Symbol:
- Desmoglein 3/PVA
- Desmoglein 3/PVA
- Antigen Name:
- CDHF6
- CDHF6
- Antigen Synonyms:
- DSG3|Desmoglein-3|130 kDa pemphigus vulgaris antigen|Cadherin family member 6|PVA
- DSG3|Desmoglein-3|130 kDa pemphigus vulgaris antigen|Cadherin family member 6|PVA
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR14101
- EPR14101
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 107533 Da
- 107533 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Concentration:
- 0.879 mg/ml
- 0.879 mg/ml
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P