Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SQSTM1/p62 Rabbit Polyclonal Antibody
Anti-SQSTM1/p62 Rabbit Polyclonal Antibody
Catalog # ABCA_AB91526-100UG
Supplier:  Abcam
New Product
Anti-SQSTM1/p62 Rabbit Polyclonal Antibody
Catalog # ABCA_AB91526-100UG
Supplier:  Abcam
Supplier Number:  AB91526-100UG
New Product

Specifications

  • Size:
    100 µg
  • Packaging:
    Vial

Specifications

About this item

Rabbit polyclonal to SQSTM1/p62.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SQSTM1/p62
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human