Anti-Prolyl Endopeptidase Rabbit Polyclonal Antibody
ABCA_AB58988-100UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:PEP
- Antigen Symbol:Prolyl Endopeptidase
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Dog
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:PREP|PE|Post-proline cleaving enzyme
- Storage Buffer:PBS, glycerol, sodium chloride
- Molecular Weight:80700 Da
- Storage Temperature:Store at −20 °C. Stable for 12 months at −20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human PREP. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to Prolyl Endopeptidase.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Prolyl Endopeptidase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog