Anti-NrCAM Mouse Monoclonal Antibody [clone: S364-51]
ABCA_AB186247-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:KIAA0343
- Antigen Symbol:NrCAM
- Clonality:Monoclonal
- Clone:S364-51
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Neuronal cell adhesion molecule|Nr-CAM|Neuronal surface protein Bravo|NgCAM-related cell adhesion molecule|hBravo|Ng-CAM-related
- Molecular Weight:130668 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Mouse Nrcam aa 1-850. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF
- Purification:Protein G affinity purification
- Packaging:Vial
Mouse monoclonal [S364-51] to NrCAM.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: NrCAM
Clonality: Monoclonal
Clone: S364-51
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Rat