Anti-Pan Trk Rabbit Monoclonal Antibody [clone: EPR18413]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR18413] to Pan Trk.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Pan Trk
Clonality: Monoclonal
Clone: EPR18413
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- ABCA_AB189903-1ML
- ABCA_AB189903-100U
- Antigen Symbol:
- Pan Trk
- Pan Trk
- Antigen Name:
- TRKC
- TRKC
- Antigen Synonyms:
- NTRK3|NT-3 growth factor receptor|GP145-Neurotrophic tyrosine kinase receptor type 3|TrkC tyrosine kinase|Trk-C
- NTRK3|NT-3 growth factor receptor|GP145-Neurotrophic tyrosine kinase receptor type 3|TrkC tyrosine kinase|Trk-C
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR18413
- EPR18413
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 92801 Da
- 92801 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IP
- IP