Anti-68kDa Neurofilament/NF-L Mouse Monoclonal Antibody [clone: DA2]
ABCA_AB7255-250UL
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:NF68
- Antigen Symbol:68kDa Neurofilament/NF-L
- Clonality:Monoclonal
- Clone:DA2
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western Blot:Yes
- Size:250 μl
- Epitope:Epitope mapped to a short peptide in the C-terminal 'tail' region of the molecule within the human sequence SYYTSHVQEEQIEVE, amino acids 441-455
- Form:Liquid
- Antigen Synonyms:NFL|NEFL|Neurofilament light polypeptide|NF-L|68 kDa neurofilament protein|Neurofilament triplet L protein
- Molecular Weight:61517 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:4 °C
- Immunogen:Native Full Length Protein corresponding to Pig NEFL
- Tested Applications:ICC/IF, IHC-P
- Purification:Tissue culture supernatant
- Packaging:Vial
Mouse monoclonal [DA2] to 68kDa Neurofilament/NF-L.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: 68kDa Neurofilament/NF-L
Clonality: Monoclonal
Clone: DA2
Conjugation:
Epitope: Epitope mapped to a short peptide in the C-terminal “tail” region of the molecule within the human sequence SYYTSHVQEEQIEVE, amino acids 441-455
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse