Anti-Rsk 2 / MAPKAP Kinase 1b Rabbit Monoclonal Antibody [clone: Y82] (APC (Allophycocyanin))
ABCA_AB305491-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:ISPK1
- Antigen Symbol:Rsk 2 / MAPKAP Kinase 1b
- Clonality:Monoclonal
- Clone:Y82
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:MAPKAPK1B|RSK2|RPS6KA3|Ribosomal protein S6 kinase alpha-3|S6K-alpha-3|90 kDa ribosomal protein S6 kinase 3|Insulin-stimulated protein kinase 1|MAP kinase-activated protein kinase 1b|Ribosomal S6 kinase 2|pp90RSK2|p90-RSK 3|p90RSK3|ISPK-1|MAPK-activated protein kinase 1b|MAPKAP kinase 1b|MAPKAPK-1b|RSK-2
- Molecular Weight:83736 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at 4 °C. Store in the dark.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:Antibody labelling, Target binding affinity
- Purification:Protein A Affinity purification
- Packaging:Vial
APC Rabbit monoclonal [Y82] to Rsk 2 / MAPKAP Kinase 1b.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ITK/EMT
Clonality: Monoclonal
Clone: Y401
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: