Anti-GABA A Receptor beta 2/GABRB2 Rabbit Polyclonal Antibody
Catalog # ABCA_AB186875-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Gamma-aminobutyric acid receptor subunit beta-2
- Antigen Symbol:GABA A Receptor beta 2/GABRB2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:GABA(A) receptor subunit beta-2|GABAAR subunit beta-2|GABRB2
- Molecular Weight:59150 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C or –80 °C. Avoid freeze / thaw cycle.
- Shipping Temperature:+4 °C
- Immunogen:Recombinant Fragment Protein within Human GABRB2 aa 1-250. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to GABA A Receptor beta 2/GABRB2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GABA A Receptor beta 2/GABRB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human