Anti-Influenza A Virus Nucleoprotein Mouse Monoclonal Antibody [clone: AA5H]
Catalog # ABCA_AB20343-250UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Nucleoprotein
- Antigen Symbol:Influenza A Virus Nucleoprotein
- Clonality:Monoclonal
- Clone:AA5H
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Influenza A
- Size:250 µg
- Form:Liquid
- Antigen Synonyms:Nucleocapsid protein|Protein N|NP
- Storage Buffer:PBS
- Molecular Weight:56225 Da
- Storage Temperature:Store at 4 °C. Do not freeze.
- Concentration:7.26 mg/ml
- Shipping Temperature:4 °C
- Tested Applications:ICC/IF, IHC-P
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [AA5H] to Influenza A Virus Nucleoprotein.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Influenza A Virus Nucleoprotein
Clonality: Monoclonal
Clone: AA5H
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Influenza A