Anti-PSMA Mouse Monoclonal Antibody [clone: GCP-05]
Catalog # ABCA_AB66912-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:FOLH
- Antigen Symbol:PSMA
- Clonality:Monoclonal
- Clone:GCP-05
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Size:100 µg
- Epitope:This antibody recognizes the extracellular domain of PSMA.
- Form:Liquid
- Antigen Synonyms:NAALAD1|PSM|GIG27|FOLH1|Glutamate carboxypeptidase 2|Cell growth-inhibiting gene 27 protein|Folate hydrolase 1|Folylpoly-gamma-glutamate carboxypeptidase|Glutamate carboxypeptidase II|Membrane glutamate carboxypeptidase|N-acetylated-alpha-linked acidic dipeptidase I|Prostate-specific membrane antigen|Pteroylpoly-gamma-glutamate carboxypeptidase|FGCP|GCPII|mGCP|NAALADase I
- Storage Buffer:PBS
- Molecular Weight:84331 Da
- Storage Temperature:Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Recombinant Fragment Protein within Human FOLH1 aa 1 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [GCP-05] to PSMA.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PSMA
Clonality: Monoclonal
Clone: GCP-05
Conjugation:
Epitope: This antibody recognizes the extracellular domain of PSMA.
Host: Mouse
Isotype: IgG1
Reactivity: Human