Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Matrix protein 1 Rabbit Monoclonal Antibody [clone: HL1275]
Anti-Matrix protein 1 Rabbit Monoclonal Antibody [clone: HL1275]
Catalog # ABCA_AB306544-100U
Supplier:  Abcam
New Product
undefined
Anti-Matrix protein 1 Rabbit Monoclonal Antibody [clone: HL1275]
Catalog # ABCA_AB306544-100U
Supplier:  Abcam
Supplier Number:  AB306544-100UL
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    M1
  • Antigen Symbol:
    Matrix protein 1
  • Clonality:
    Monoclonal
  • Clone:
    HL1275
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western Blot:
    Yes
  • Size:
    100 µl
  • Form:
    Liquid
  • Antigen Synonyms:
    M
  • Storage Temperature:
    Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
  • Concentration:
    1 mg/ml
  • Shipping Temperature:
    4 °C
  • Purification:
    Protein A Affinity purification
  • Packaging:
    Vial

Specifications

About this item

Rabbit monoclonal [HL1275] to Matrix protein 1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HLA-DP
Clonality: Monoclonal
Clone: SP228
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: