Anti-Nkx2.2 Mouse Monoclonal Antibody [clone: NX2/294]
Catalog # ABCA_AB187375-500U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:NKX2B
- Antigen Symbol:Nkx2.2
- Clonality:Monoclonal
- Clone:NX2/294
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Size:500 µl
- Form:Liquid
- Antigen Synonyms:NKX2-2|Homeobox protein Nkx-2.2|Homeobox protein NK-2 homolog B
- Molecular Weight:30133 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:0.2 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human NKX2-2
- Tested Applications:IHC-P
- Purification:Protein A/G affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [NX2/294] to Nkx2.2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Nkx2.2
Clonality: Monoclonal
Clone: NX2/294
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Rat